Surface protection sheets, BOLA Sheets PTFE or FEP
Supplier: BOHLENDER
Certificates
About this item
Protection sheets made from PTFE or FEP.
- PTFE rolls in white
- FEP rolls in transparent, gastight and non-porous
- Temperature resistant from −200 to +250 °C
For use as table pad or lining drawers. Also suitable for using as slideway or for insulation.
Delivery: Delivered on rolls.