订单输入
China
ContactUsLinkComponent
叔丁醇锂 2.2 M 在四氢呋喃中, AcroSeal®
Corrosive
Flammable
Irritant
Health
叔丁醇锂 2.2 M 在四氢呋喃中, AcroSeal®
Certificates
undefined
Corrosive
Flammable
Irritant
Health
叔丁醇锂 2.2 M 在四氢呋喃中, AcroSeal®
Specification Test Results
  • Appearance:
    Clear yellow to brown Solution
  • Strength:
    19 to 21 %
  • Impurity:
    ≤0.2 % LiOH + Li2CO3 (as LiOH)
Specification Test Results