
Curcumin 98+% (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:458-37-7
:
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365
:368.39 g/mol
Melting Pt:183 °C
EINECS:207-280-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange Crystalline powder
- Infrared spectrum:Conforms
- UV spectrophotometry:≥98.0 %