Order Entry
China
ContactUsLinkComponent
Alcohol, anhydrous 94.0-96.0% (v/v), BAKER ANALYZED® ACS (denatured with methanol and isopropanol), J.T.Baker®
Flammable
Health
Irritant
Alcohol, anhydrous 94.0-96.0% (v/v), BAKER ANALYZED® ACS (denatured with methanol and isopropanol), J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Health
Irritant
Alcohol, anhydrous 94.0-96.0% (v/v), BAKER ANALYZED® ACS (denatured with methanol and isopropanol), J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    94.0 - 96.0 %
  • For Laboratory,Research,or Manufacturing Use:
    4.0 - 6.0 %
  • Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v):
    94.0 - 96.0 %
  • Isopropyl Alcohol (2-propanol)(v/v):
    4.0 - 6.0 %
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 0.001 %
  • Specific Gravity at 25°C:
    0.785 - 0.795
  • Water (H₂O):
    ≤ 0.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number