


Lithium-tert-butanolate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1907-33-1
:
Formula:C₄H₉LiO
MDL Number:MFCD00050479
:80.06 g/mol
Density:0.89 g/cm³
Storage Temperature:Ambient
EINECS:217-611-5
UN:2925
ADR:4.1,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Titration with HCl:≥98.5 %
- Impurity:≤1 % (LiOH + Li2CO3)