L(+)-Ascorbic acid 99.0-100.5%, fine powder USP 200-325 Mesh, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
CAS Number:50-81-7
:
Formula:C₆H₈O₆
MDL Number:MFCD00064328
Boiling Pt:553
Melting Pt:190…192 °C
Density:1.65 g/cm³ (20 °C)
Molecular Weight:176.13 g/mol
Merck Index:13,00837
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets J.P. Chemical Specifications:99.0 - 100.5%
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:20.5 - 21.5°
- Bulk Pharmaceutical Chemical:≤0.1%
- USP - Assay (C₆H₈O₆):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):20.5 - 21.5 Degree
- USP - Residue on Ignition:≤0.1 %
- Mesh - Thru U.S. No. 100 Sieve:≥100 %
- Mesh - Thru U.S. No. 200 Sieve:≥95 %
- JP - Assay (C₆H₈O₆) (dried basis):99.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Optical Rotation [Å]²º^D (+):20.5 - 21.5 Degree
- JP - pH (1 in 20):2.2 - 2.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Loss on Drying:≤0.20 %
- JP - Residue on Ignition:≤0.10 %