Starch indicator solution, MDB
Catalog # HACH34932
Supplier: HACH
CAS Number:
Specifications
- Pk:100 mL
- Description:Starch indicator solution
Specifications
About this item
Oxidation-reduction indicator for the analysis of BOD, total chlorine, chromium, cyanide, dissolved oxygen and sulfite.
- Marked dropping bottle (MDB)