
9-Aminoacridine hemihydrate 98%
Catalog # ACRO134410010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
- Specific optical rotation:+85° to +95° (20°C, 589 nm) (c=0.5, methanol)
Specification Test Results
About this item
:
:203.24 g/mol
Melting Pt:230…235 °C
EINECS:201-995-6
:GHS07
Storage Temperature:Ambient
CAS Number:65944-23-2
Formula:C₁₃H₁₀N₂·0.5H₂O
MDL Number:MFCD28040496