Order Entry
China
ContactUsLinkComponent
Pyridine, anhydrous ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS, ISO analytical reagent, for Karl Fischer titration
Flammable
Irritant
Pyridine, anhydrous ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS, ISO analytical reagent, for Karl Fischer titration
Catalog # VWRC27198.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Pyridine, anhydrous ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS, ISO analytical reagent, for Karl Fischer titration
Catalog # VWRC27198.298
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Assay (GC):
    Min. 99.7 %
  • Acidity or alkalinity:
    Max. 0.200 meq/g
  • Water:
    Max. 0.0200 %
  • Evaporation residue:
    Max. 0.0020 %
  • Reducing substances:
    Max. 0.0005 %
  • Piperidine:
    Max. 0.0100 %
  • 2-Methylpyridine:
    Max. 0.20 %
  • NH4 (Ammonium):
    Max. 0.0010 %
  • Cl (Chloride):
    Max. 0.0005 %
  • SO4 (Sulphate):
    Max. 0.0005 %
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.05 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Ni (Nickel):
    Max. 0.01 ppm
  • Pb (Lead):
    Max. 0.01 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.05 ppm

Specification Test Results

About this item

 :
Merck Index:13,08060
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
 :79.1 g/mol
 :GHS02
 :GHS07
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732