

Pyridine, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Merck Index:13,08060
Boiling Pt:115…116 °C (760 mmHg)
Melting Pt:–42 °C
Density:0.978 g/cm³
Flash Pt:17 °C
UN:1282
ADR:3,II
:79.1 g/mol
EINECS:203-809-9
:GHS07
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.4 %
- Residue after evaporation:≤0.001 %