Calcium chloride, dry ≥90%, granules 2-6 mm, purified
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.174 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 90 %
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC22316.261 spec_VWRC22316.261 spec_VWRC22316.261 spec_VWRC22316.363 spec_VWRC22316.363
Product specification
More
VWR® for Drying and Filtration