
Lithium sulphate, anhydrous 99%, extra pure
Catalog # ACRO218322500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Appearance:White Powder
- Water:≤1 % (K.F.)
- Titration after Ion exchange:≥98.5 %
- Insoluble matter:≤0.01 %
Specification Test Results
About this item
:
:109.95 g/mol
Merck Index:12,05569
Boiling Pt:1377 °C (1013 hPa)
Melting Pt:859 °C
Density:2.22 g/cm³
Storage Temperature:Ambient
EINECS:233-820-4
:GHS07
CAS Number:10377-48-7
Formula:Li₂O₄S
MDL Number:MFCD00011086