Order Entry
China
ContactUsLinkComponent
 
Sodium thiosulphate, anhydrous 99%
Catalog # ALFAA17629.36
CAS Number:  
Sodium thiosulphate, anhydrous 99%
Catalog # ALFAA17629.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :158.11 g/mol
Merck Index:14,8694
Boiling Pt:100 °C (1013 hPa)
Melting Pt:48 °C
Density:1.667
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499