

Sodium nitrate 99+% for analysis
Catalog # ACRO205960010
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- pH:5.5 to 8.3 (5 % at 25°C)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤10 ppm
- Nitrite (NO2):≤10 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤30 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤3 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
About this item
:
:84.99 g/mol
Merck Index:13,08720
Boiling Pt:380 °C (1013 hPa)
Melting Pt:306 °C
Density:2,26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-554-3
UN:1498
ADR:5.1,III
REACH:01-2119488221-41
:GHS07
:GHS03
CAS Number:7631-99-4
Formula:NaNO₃
MDL Number:MFCD00011119