Potassium nitrate ≥99% ACS
Jump to:
Product Details & Documents
Synonyms: Nitrate of Potash
- CAS Number:7757-79-1
- Formula:KNO₃
- MDL Number:MFCD00011409
- Molecular Weight:101.1 g/mol
- Merck Index:13,07733
- Melting Pt:334 °C
- Density:2.109 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-818-8
- UN:1486
- ADR:5.1,III
- REACH:01-2119488224-35
Documents
Specifications
Specification Test Results
- Calcium:<0.005%
- Chloride:≤0.002%
- Heavy Metals (as Pb):≤0.0005%
- Insolubles:≤0.005%
- Iodate:≤0.0005%
- Iron:≤0.0003%
- Magnesium:≤0.002%
- Nitrite:≤0.001%
- pH (5%, Water) @25 °C:4.5 – 8.5
- Phosphate:≤0.0005%
- Purity:≥99%
- Sodium:≤0.005%
- Sulphate:≤0.003%



