Order Entry
China
ContactUsLinkComponent
 
 
Lithium fluoride 97% for analysis
 
undefined
 
Lithium fluoride 97% for analysis

 

  • Appearance:
    White to off-white Powder
  • Assay:
    ≥96.0 %
  • Loss on drying:
    0.4 % (200°C) (typical)
  • Chloride (Cl):
    ≤0.07 %
  • Carbonate (CO3):
    ≤1 %
  • Phosphate (PO4):
    ≤0.04 %
  • Sulfate (SO4):
    ≤1 %
  • Calcium (Ca):
    ≤0.08 %
  • Iron (Fe):
    ≤0.03 %
  • Magnesium (Mg):
    ≤0.02 %
  • Potassium (K):
    ≤0.07 %
  • Sodium (Na):
    ≤0.4 %