Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:205-633-8
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification:Passes Test
- USP - Limit of Sulfur Compounds:≤ 0.015 %
- USP - Loss on Drying:≤ 0.25 %
- USP - Insoluble Matter:Passes Test
- USP - Normal Carbonate:Passes Test
- USP - Limit of Ammonia:≤ 20 ppm
- USP - Chloride (Cl):≤ 0.015 %
- USP - Arsenic (As):≤ 2 ppm
- USP - Assay (NaHCO₃) (dried basis):99.0 - 100.5 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Assay (NaHCO₃):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Carbonate (CO₃):Passes Test
- EP/BP - Chloride (Cl):≤ 150 ppm
- EP/BP - Sulfate (SO₄):≤ 150 ppm
- EP/BP - Ammonium (NH₄):≤ 20 ppm
- EP/BP - Calcium (Ca):≤ 100 ppm
- EP/BP - Iron (Fe):≤ 20 ppm
- JP - Assay (NaHCO₃):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - pH (1 in 20):7.9 - 8.4
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.040 %
- JP - Carbonate (CO₃):Passes Test
- JP - Ammonium (NH₄):Passes Test
- JP - Heavy Metals (as Pb):≤ 5 ppm
- JP - Arsenic (As):≤ 2 ppm
- No Class 1,2,3 or other solvents are used or produced in the:= 5 ppm
- manufacturing or purification of the product.:= 2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR3510-05 spec_BAKR3510-07 spec_BAKR3510-09