Order Entry
China
ContactUsLinkComponent
 
 
Lithium carbonate 99.999% (trace metals basis), extra pure
  ACRO193360100
 :  
undefined
 
Lithium carbonate 99.999% (trace metals basis), extra pure
  ACRO193360100
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White Powder
  • Total trace metal impurities:
    ≤0.001 %

 

 

 :
 :73.89 g/mol
 :13,05549
 :720 °C
 :2,11 g/cm³ (20 °C)
 :209-062-5
 :GHS07
 :Ambient
 :554-13-2
 :Li₂CO₃
 :MFCD00011084