Order Entry
China
ContactUsLinkComponent
Lithium bromide 99.999% (trace metals basis), extra pure
 
Lithium bromide 99.999% (trace metals basis), extra pure
  ACRO212530250
 :  
undefined
 
Lithium bromide 99.999% (trace metals basis), extra pure
  ACRO212530250
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to off-white Powder or crystals and/or chunks
  • Total trace metal impurities:
    ≤10 ppm

 

 

 :
 :86.84 g/mol
 :13,05548
 :1265 °C (760 mmHg)
 :550 °C
 :3.46 g/cm³
 :Ambient
 :231-439-8
 :GHS07
 :7550-35-8
 :23303-71-1
 :LiBr
 :MFCD00011077