Order Entry
China
ContactUsLinkComponent
 
 
Potassium bromide, Uvasol® for IR spectrophotometry, Supelco®
  MERC1.04907.0100
 :  
undefined
 
Potassium bromide, Uvasol® for IR spectrophotometry, Supelco®
  MERC1.04907.0100
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Solubility:
    650 g/L (20 °C)
  • Melting Point:
    730 °C
  • Molar Mass:
    119.00 g/mol
  • Bulk Density:
    900 - 1000 kg/m³
  • Boiling Point:
    1435 °C
  • Vapor Pressure:
    1.3 hPa (795 °C)
  • Density:
    2.75 g/cm³ (20 °C)
  • pH:
    5.5 - 8.5 (50 g/L, H₂O, 20 °C)

 

 

 :
 :13,07701
 :1435 °C (1013 hPa)
 :730 °C
 :2.75 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :119 g/mol
 :Ambient
 :231-830-3
 :GHS07
 :7758-02-3
 :KBr
 :MFCD00011358