Order Entry
China
ContactUsLinkComponent
Potassium bromide 99+% for IR spectrophotometry
Irritant
Potassium bromide 99+% for IR spectrophotometry
Catalog # ACRO206391000
CAS Number:  
undefined
Irritant
Potassium bromide 99+% for IR spectrophotometry
Catalog # ACRO206391000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥99.0 %
  • Additional info:
    Application test: Passes test
  • Absorbance:
    (2 g pressed disk, 0.5 cm layer thickness)

Specification Test Results

About this item

 :
Merck Index:13,07701
Boiling Pt:1435 °C (760 mmHg)
Melting Pt:734 °C
Density:2,75 g/cm³ (20 °C)
 :119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
 :GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358