Order Entry
China
ContactUsLinkComponent
Brij® 35 30% in aqueous solution, Pierce™
Brij® 35 30% in aqueous solution, Pierce™
Catalog # PIER20150
CAS Number:  
undefined
Brij® 35 30% in aqueous solution, Pierce™
Catalog # PIER20150
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    950 ml
  • Seq:
    2845

Specifications

About this item

 :
 :1199.56 g/mol
EINECS:500-002-6
CAS Number:9002-92-0
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891