
Lithium trifluoromethanesulphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33454-82-9
:
Formula:CF₃LiO₃S
MDL Number:MFCD00013227
:156.01 g/mol
Melting Pt:423 °C
EINECS:251-528-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Assay:4.34 to 4.56 % (Li)