TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29915-38-6
:
Formula:C₇H₁₇NO₆S
MDL Number:MFCD00007538
:243.28 g/mol
Melting Pt:240 °C (dec.)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:249-954-1
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- UV:(0.1 M in water)
- pH:5 to 6.5 (1 % in water at 25°C)
- Titration with NaOH:≥99 %