



Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Catalog # ACRO406025000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.02 % (Coulometric)
- Boiling point:79 to 81 °C
- Density:(20/20 °C) 0.779 to 0.780
- GC:≥99.0 %
- UV:at 210 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Substance darkened by H2SO4:passes test
Specification Test Results
About this item
:
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (760 mmHg)
Melting Pt:6.5 °C
Density:0.77 g/cm³
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814