Cyclohexanone ≥99.0% analytical reagent
Glass bottle, 1 L
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Catalog # VWRC23236.294
Supplier: VWR Chemicals
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155.6 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0.9478 g/cm³ (20 °C)
- Flash Pt:43 °C
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
P20_VWRC23236.294_SPEC_EN.pdf spec_VWRC23236.294 spec_VWRC23236.294 spec_VWRC23236.460
Product specification
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Acidity:Max. 0.005 meq/g
- Boiling point:154.5 - 156.5 °C
- Density (20/4):0.946 - 0.948
- Refractive index (20°C):1.449 - 1.451
- Viscosity (25°C):1.92 - 2.2 mPa·s
- Cyclohexanol:Max. 0.5 %
- Water:Max. 0.2 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:VWRC23236.294
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



