Sorbic acid
Glass bottle for solids, 10 mg
Catalog # EDQMS0950000
Supplier: EDQM
Jump to:
Product Details & Documents
同义词: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
- CAS号:110-44-1
- 化学式:CH₃CH=CHCH=CHCOOH
- MDL 编号:MFCD00002703
- Molecular Weight:112.13 g/mol
- 默克索引:13,08793
- 沸点:228 °C (1013 hPa)
- 熔点:132…135 °C
- 密度:1.204 g/cm³ (20 °C)
- 闪点:127 °C (closed cup)
- 储存温度:Ambient
- EINECS:203-768-7
规格
规格
- Pack type:Glass bottle for solids
- Pk:10 mg



