Urea 7.8-8.2 M in aqueous solution, Ultra Pure Grade
VWRCE720-250ML
:
- Pack type:Plastic bottle
- Pk:250 mL
- Seq:2590
- Environmentally Preferable:
- Appearance:Clear and colorless liquid
- Urea:7.8 Molar - 8.2 Molar
: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
:
:60.06 g/mol
:197
:132.7
:Ambient
:200-315-5
:57-13-6
:NH₂CONH₂
:MFCD00008022
:13,09935