
1,2-Dibromo-1,2-diphenylethane 96%
Catalog # ACRO147950050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow to light orange Powder
- Infrared spectrum:Conforms
- Melting point:234 to 241 °C
- HPLC:≥95 %
Specification Test Results
About this item
:
:340.06 g/mol
Melting Pt:235…241 °C (dec.)
Storage Temperature:Ambient
EINECS:227-324-7
UN:3261
ADR:8,III
:GHS05
CAS Number:5789-30-0
Formula:C₆H₅(CHBr)₂C₆H₅
MDL Number:MFCD00000137