Order Entry
China
ContactUsLinkComponent
Toluene ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
 :  Avantor
undefined
Toluene ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
 :  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • For Laboratory,Research,or Manufacturing Use:
    ≥ 99.7 %
  • Assay (by GC, corrected for water):
    ≥ 99.7 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤ 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:
    ≤ 0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm:
    ≤ 0.40
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤ 285
  • Residue after Evaporation:
    ≤ 3.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤ 0.02 %