


Toluene ≥99.7% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
: Avantor
:108-88-3
:
:C₆H₅CH₃
:MFCD00008512
:92.14 g/mol
:13,09607
:110.6 °C (1013 hPa)
:–95 °C
:0.867 g/cm³ (20 °C)
:Ambient
:203-625-9
:1294
:3,II
:Packaged under inert gas.
:4 °C
:05-2114615130-69
:GHS02
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- For Laboratory,Research,or Manufacturing Use:= 10
- Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤10
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- Assay (C₆H₅CH₃) (by GC, corrected for water):≥99.7 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤0.03 %