Order Entry
China
Orders LinkContactUsLinkComponent
Cumene 98%

Cumene 98%

Flammable
Irritant
Health
Environment
Cumene 98%
Certificates
Cumene 98%

2 Options Available Below

Product Details & Documents
Synonyms:  2-Phenylpropane, Isopropylbenzene
  • CAS Number:98-82-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008881
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,02644
  • Boiling Pt:151 °C (1013 hPa)
  • Melting Pt:–96 °C
  • Density:0.862 g/cm³
  • Flash Pt:31 °C
  • Storage Temperature:Ambient
  • EINECS:202-704-5
  • UN:1918
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4890 to 1.4930 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 升
    1 * 1 升

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 升
    1 * 2,5 升