Order Entry
China
ContactUsLinkComponent
Overhead stirrer tool, radial stirrer
Overhead stirrer tool, radial stirrer
Catalog # BOCH9030
Supplier:  BOCHEM
undefined
Overhead stirrer tool, radial stirrer
Catalog # BOCH9030
Supplier:  BOCHEM

Specifications

  • Environmentally Preferable:
  • Pk:
    1
  • Shaft Ø:
    7 mm
  • Shaft length:
    400 mm
  • Stirrer Ø:
    50 mm

Specifications

About this item

Radial stirrer made of stainless steel.

  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low and medium viscous media
  • Creates radial flow
  • Ideal for aeration of liquids