Overhead stirrer tools, paddle stirrers
Supplier: BOCHEM
Certificates
About this item
Paddle stirrers made of stainless steel, with six holes.
- Ideal for stirring liquids at middle speed range
- Suitable for stirring low to medium viscous media
- Creates radial flow