Stirrers, propeller, with two paddles
Supplier: BOCHEM
Certificates
About this item
Propeller stirrer made of stainless steel with two fixed paddles.
- Mixing of media with low and middle viscosity
- Radial flow
Propeller stirrer made of stainless steel with two fixed paddles.