Stirrers, propeller, with four paddles
Supplier: BOCHEM
Certificates
About this item
Propeller stirrer made of stainless steel with four paddles.
- Mixing of media with low and middle viscosity
- Ideal for homogenisation
- Axial flow
Propeller stirrer made of stainless steel with four paddles.