Order Entry
China
ContactUsLinkComponent
Labels, seal-it
Labels, seal-it
Catalog # BURK5399-0003
Supplier:  BURKLE
undefined
Labels, seal-it
Catalog # BURK5399-0003
Supplier:  BURKLE
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Colour:
    White/red
  • Description:
    Security seal for retention samples and tamper-proof packs
  • Dimensions:
    178×30 mm
  • Pk:
    1 Roll
Specifications
About this item
Retention samples are kept to provide a sample for comparison at a later date. The unambiguous proof that the retention sample is identical to the original sample is only possible with clearly marked and sealed containers. However, sealing is frequently not possible unless eyelets are present. seal-it provides the solution! The seal-it self-adhesive label sticks to all conventional containers. Thanks to its unbelievable adhesive power it cannot be removed. If a container opening is closed with seal-it, then the container cannot be opened without destroying the seal-it label. seal-it is additionally provided with several cross-perforations which immediately tear when manipulated like a motorcar tax disc. seal-it can be used to securely seal bottles, sample bags, sacks, specimen bags, envelopes, and more.
  • Clear identification via numbers in sequence
  • Integrated control label with the same number, for accompanying documents
  • Tear-off perforation for each label
  • Can be written on with all conventional writing tools
  • The aluminium foil adapts seal-it to any container shape