Order Entry
China
Orders LinkContactUsLinkComponent
Mini immersion cylinder

Mini immersion cylinder

Mini immersion cylinder, Stainless steel V2A (1.4301), 50 ml, 180 mm, 32 mm
Catalog # BURK5365-5000
Supplier:   BURKLE
Mini immersion cylinder
Catalog # BURK5365-5000
Supplier:  BURKLE
Mini immersion cylinder
Mini immersion cylinder

Mini immersion cylinder, Stainless steel V2A (1.4301), 50 ml, 180 mm, 32 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Mini immersion cylinder for sampling liquids in narrow places where access is difficult.

  • Screw-off top enables easy cleaning
  • Tested for use in the Ex protection zone 0
  • Explosion Categories IIA and IIB

Can also be used in curved dipping tubes and barrels with narrow openings.

Ordering information: Please order the appropriate lowering cable at the same time.

Specifications
  • Ø:
    32 mm
  • Capacity:
    50 ml
  • Description:
    Mini immersion cylinder
  • Length:
    180 mm
  • Material:
    Stainless steel V2A (1.4301)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.