Order Entry
China
ContactUsLinkComponent
 
Irritant
Health
N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®
Catalog # ACRO375231000
CAS Number:  
undefined
Irritant
Health
N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®
Catalog # ACRO375231000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.4 %

Specification Test Results

About this item

 :
 :87.12 g/mol
Merck Index:13,03254
Boiling Pt:164…166 °C (760 mmHg)
Melting Pt:–20 °C
Density:0.937 g/cm³
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686