About this item
PVC, transparent. Flexible and transparent hoses manufactured in atoxic material.
- Suitable for liquids transfer
- Cadmium free
- Temperature resistance: -5 to +50 °C
Shore hardness: 65 ±5