

2-Iodobenzoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88-67-5
:
Formula:C₇H₅IO₂
MDL Number:MFCD00002419
:248.02 g/mol
Merck Index:13,05052
Boiling Pt:314 °C (1013 hPa)
Melting Pt:161…163 °C
Density:2,25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-850-7
:GHS07
:GHS05
Specification Test Results
- Appearance:White to off-white to beige Powder
- Infrared spectrum:Conforms
- Melting point:160 to 164 °C
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
- Calcium (Ca):≤0.003 %
- Magnesium (Mg):≤0.005 %
- Potassium (K):≤0.01 %
- Insoluble matter:≤0.005 %