Order Entry
China
ContactUsLinkComponent
RNA collection tubes, BD PAXgene™
RNA collection tubes, BD PAXgene™
Catalog # BDAM762165
Supplier:  BD Medical
RNA collection tubes, BD PAXgene™
Catalog # BDAM762165
Supplier:  BD Medical

Specifications

  • Closure type:
    BD Hemogard™
  • Ø:
    16 mm
  • Volume:
    2,5 ml
  • Description:
    Blood Collection tube, PAXgene®
  • Length:
    100 mm
  • Material:
    PET
  • Cap colour:
    Red
  • Pk:
    100
  • Additive:
    Proprietary 6,9 ml
  • Label type:
    Paper label

Specifications

About this item

The tube contains a proprietary reagent that immediately stabilises all total RNA, including miRNA.

  • Enables accurate detection and quantitation of gene transcripts
  • Minimises ex vivo blood RNA degradation or gene induction

Stabilises RNA for three days at room temperature (18 to 25 °C) and five days at 4 °C. Paxgene™ tubes can be frozen at –20 °C or –70 °C for long term storage.