Order Entry
China
Orders LinkContactUsLinkComponent
Specimen container, disposable, with lid
Specimen container, disposable, with lid
128 ml, Yellow
Catalog # SIMPC570-12
Supplier:   SIMPORT SCIENTIFIC
Specimen container, disposable, with lid
Catalog # SIMPC570-12
Supplier:  SIMPORT SCIENTIFIC
Specimen container, disposable, with lid
Specimen container, disposable, with lid
128 ml, Yellow
Price
Inquire For Price
Log in for availability
1 * 500 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Graduated container (PP) with screw cap (PE). Heavy-duty thick wall construction of both container and lid ensures a positive leakproof seal.
  • Wide base design for stable reliable use
  • Molded-in graduations up to 128 ml
  • Non-sterile
The drip ring on the container reduces the chances of contamination.
Packaging: Packaged in bags of 100 stacked by 10's. Yellow caps packed separately in bags of 100.
Specifications
  • Capacity:
    128 ml
  • Cap colour:
    Yellow