- Capacity:50 ml
- Colour:Green
- Material:PP
- Pk:72
The SputEm™ Sputum Collection System offers an ideal method to collect, carry, and process biological samples.
- Available in a light green color, three narrow vertical windows allow the contents of the tube to be discreetly viewed
- A wide base ensures great stability and prevents tipping
- The large collection funnel is made in such a way that specimens fall directly into the graduated centrifuge tube and do not contaminate the outside threads
- The centrifuge tube screw cap is kept sterile at all times under the snap cap on top of the funnel
A patient label is already affixed on top of the unit and can easily be transferred to the tube before leaving for the laboratory. The base can be used as a support during transit.
The unit features a base which incorporates a removable, sterile, 50mL graduated polypropylene conical tube that can withstand centrifugation up to 5000 rpm or 3000×g.