Graduated pipettes, Class AS, total and partial delivery, DURAN®
Supplier: DWK LIFE SCIENCES
Certificates
About this item
Soda-lime glass (AR-Glas®).
- Batch certificate and certificate of conformity
- Printed Retrace Code (batch identification)
- For cotton plugs
- Marks and inscription in brown with ring graduations
- Numbering: Zero at top
- Graduated to tip (total delivery, type 3)
Complies to DIN ISO 835.
Colour code complies to DIN 12621.
Length: 360 mm for items with 25 ml or more 450 mm.