
(1R,2R)-(+)-1,2-Diphenylethylenediamine 99%, 99% ee
Catalog # ACRO294682500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:81 to 84 °C
- GC:≥98.5 %
- Enantiomeric excess:≥98.5 %
- Specific optical rotation:+102° to +106° (25 °C, 589 nm) (c=1.1, MeOH)
Specification Test Results
About this item
:
:212.29 g/mol
Melting Pt:81…84 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:35132-20-8
Formula:C₁₄H₁₆N₂
MDL Number:MFCD00082769