
Trilithium citrate tetrahydrate 98%, pure
ACRO199855000
:
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤24 %
- Titration with HClO4:≥97.5 % (On anhydrous substance)
:
:281.99 g/mol
:112 °C (dec.)
:213-045-8
:GHS07
:6080-58-6
:C₆H₅Li₃O₇·4H₂O
:MFCD00150030