Order Entry
China
ContactUsLinkComponent
2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%
 
2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%
  ACRO168070050
 :  
undefined
 
2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%
  ACRO168070050
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to cream to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    248 to 252 °C
  • Titration with HClO4:
    98.5 to 101.5 %

 

 

 :
 :312.33 g/mol
 :248…252 °C
 :222-965-9
 :GHS07
 :Ambient
 :3682-35-7
 :C₁₈H₁₂N₆
 :MFCD00006045