Top tilt measures
Supplier: BRAND
Certificates
About this item
Dispenser head made of borosilicate glass 3.3, with inscriptions in green enamel.
- Calibrated to deliver (TD, Ex)
- Excellent chemical resistance
- Excellent good temperature resistance
Automatic dispenser particularly suited for dispensing suspensions.
Ordering information: Reservoir bottles available on request.