Order Entry
China
ContactUsLinkComponent
 
4,4-Dimethyl-2-pentanone 99%
  ACRO159710050
 :  
undefined
4,4-Dimethyl-2-pentanone 99%
  ACRO159710050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4025 to 1.4045 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

 : 4,4-Dimethylpentan-2-one
 :
 :114.19 g/mol
 :125…130 °C (760 mmHg)
 :–64 °C
 :0.804 g/cm³
 :18 °C
 :Ambient
 :209-685-2
 :1224
 :3,II
 :GHS02
 :590-50-1
 :C₇H₁₄O
 :MFCD00008854